9,9-Dimethylfluorene-2-boronic acid pinacol ester 95%

Lieferant: Thermo Fisher Scientific

H51950.03 H51950.06
H51950.03EA 48.7 EUR
H51950.03 H51950.06
9,9-Dimethylfluorene-2-boronic acid pinacol ester 95%
9,9-Dimethylfluorene-2-boronic acid pinacol ester

Purity: 95% CAS No.: 569343-09-5 Molecular Formula: C21H25BO2 Formula Weight: 320.24 MDL No.: MFCD08704229

Formel: C₂₁H₂₅BO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08704229
CAS-Nummer: 569343-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR