Boc-Dap(Fmoc)-OH 98%

Lieferant: Thermo Fisher Scientific

H51964.03 H51964.06
H51964.03EA 108 EUR
H51964.03 H51964.06
Boc-Dap(Fmoc)-OH 98%
Boc-Dap(Fmoc)-OH

Purity: 98% CAS No.: 122235-70-5 Molecular Formula: C23H26N2O6 Formula Weight: 426.47 Beilstein Reference: 4771385 MDL No.: MFCD00235897

Formel: C₂₃H₂₆N₂O₆
Molecular Weight: 426.469 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00235897
CAS-Nummer: 122235-70-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR