Boc-D-Tic-OH 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Boc-D-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

H51965.03 H51965.06
H51965.03EA 75.2 EUR
H51965.03 H51965.06
Boc-D-Tic-OH 98%
Boc-D-Tic-OH

Purity: 98% CAS No.: 115962-35-1 Molecular Formula: C15H19NO4 Formula Weight: 277.32 MDL No.: MFCD00143818

Formel: C₁₅H₁₉NO₄
Molecular Weight: 277,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00143818
CAS-Nummer: 115962-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR