(2S)-2-({[(2-Methyl-2-propanyl)oxy]carbonyl}amino)-4-phenylbutansäure 98%

Lieferant: Thermo Scientific

Synonyme: N-Boc-L-homophenylalanine, (S)-2-(tert-Butoxycarbonylamino)-4-phenylbutyric Acid, Boc-Homophe-OH, (S)-2-(Boc-amino)-4-phenylbutyric Acid

H51966.03 H51966.06
H51966.03EA 80.6 EUR
H51966.03 H51966.06
(2S)-2-({[(2-Methyl-2-propanyl)oxy]carbonyl}amino)-4-phenylbutansäure 98%
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid

Purity: 98% CAS No.: 100564-78-1 Molecular Formula: C15H21NO4 Formula Weight: 279.34 Melting Point: 76-80° Beilstein Reference: 3653506 Optical Rotation: +6.5° (c=2 in ethanol) MDL No.: MFCD00076904

Formel: C₁₅H₂₁NO₄
Molecular Weight: 279,34 g/mol
Schmelzpunkt: 76…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00076904
CAS-Nummer: 100564-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR