Fmoc-Tic-OH 95%

Lieferant: Thermo Fisher Scientific

Synonyme: Fmoc-L-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, (S)-2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

H51975.03 H51975.06
H51975.03EA 104 EUR
H51975.03 H51975.06
Fmoc-Tic-OH 95%
Fmoc-Tic-OH

Purity: 95% CAS No.: 136030-33-6 Molecular Formula: C25H21NO4 Formula Weight: 399.45 Beilstein Reference: 4827574 Optical Rotation: +26.0° (c=1 in methanol) MDL No.: MFCD00144368

Formel: C₂₅H₂₁NO₄
Molecular Weight: 399.446 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00144368
CAS-Nummer: 136030-33-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR