Fmoc-4-iodo-L-phenylalanine 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-p-iodo-Phe-OH, Fmoc-Phe(4-I)-OH
H51977.03
H51977.06
H51977.03EA
125
EUR
H51977.03
H51977.06
Fmoc-4-iodo-L-phenylalanine 95%
Fmoc-4-iodo-L-phenylalanine
Purity: 95%
CAS No.: 82565-68-2
Molecular Formula: C24H20INO4
Formula Weight: 513.33
Melting Point: 213-217°
Optical Rotation: -10° (c=1 in DMF)
MDL No.: MFCD00672340
Formel:
C₂₄H₂₀INO₄ Schmelzpunkt: 213…217 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672340 CAS-Nummer: 82565-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...