N-Boc-4-methyl-L-phenylalanine 95%

Lieferant: Thermo Scientific

Synonyme: Boc-p-Me-Phe-OH, Boc-Phe(4-Me)-OH

H51983.06 H51983.03
H51983.06EA 507 EUR
H51983.06 H51983.03
N-Boc-4-methyl-L-phenylalanine 95%
N-Boc-4-methyl-L-phenylalanine

Purity: 95% CAS No.: 80102-26-7 Molecular Formula: C15H21NO4 Formula Weight: 279.34 Melting Point: 84-88° Beilstein Reference: 5381557 Optical Rotation: +16° (c=1 in methanol) MDL No.: MFCD01317027

Formel: C₁₅H₂₁NO₄
Molecular Weight: 279,336 g/mol
Schmelzpunkt: 84…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01317027
CAS-Nummer: 80102-26-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR