(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid 95%

Lieferant: Thermo Fisher Scientific

Synonyme: Boc-Phe(4-NO2)-OH, Boc-Phe(4-NO₂)-OH, N-Boc-4-nitro-L-phenylalanine, Boc-p-nitro-Phe-OH

H51986.MD H51986.03 H51986.06
H51986.MDEA 26.4 EUR
H51986.MD H51986.03 H51986.06
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid 95%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid

Purity: 95% CAS No.: 33305-77-0 Molecular Formula: C14H18N2O6 Formula Weight: 310.31 Beilstein Reference: 2820671 Optical Rotation: +8° (c=1 in methanol) MDL No.: MFCD00038128

Formel: C₁₄H₁₈N₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00038128
CAS-Nummer: 33305-77-0
EINECS: 251-450-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR