Fmoc-Dap(Boc)-OH 95%
Lieferant: Thermo Scientific
Synonyme:
Fmoc-(N-beta-boc)-L-alpha,beta-diaminopropionic acid, 3-(tert-Butoxycarbonyl)amino-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-alanine, (S)-3-(tert-Butoxycarbonylamino)-2-[(9H-fluoren-9-ylmethoxy)carbonylamino]propionic acid, Nα-Fmoc-Nβ-Boc-L-2,3-diaminopropionic acid, (S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 3-(Boc-amino)-N-Fmoc-L-alanine
H51987.03
H51987.06
H51987.03EA
124
EUR
H51987.03
H51987.06
Fmoc-Dap(Boc)-OH 95%
Fmoc-Dap(Boc)-OH
Purity: 95%
CAS No.: 162558-25-0
Molecular Formula: C23H26N2O6
Formula Weight: 426.47
Storage Temperature: 2°C to 8°C
Beilstein Reference: 7057792
MDL No.: MFCD00235896
Formel:
C₂₃H₂₆N₂O₆ Molecular Weight: 426,47 g/mol |
MDL:
MFCD00235896 CAS-Nummer: 162558-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...