Boc-Dab(Fmoc)-OH 95%

Lieferant: Thermo Scientific

H51989.03 H51989.06
H51989.03EA 81.5 EUR
H51989.03 H51989.06
Boc-Dab(Fmoc)-OH 95%
Boc-Dab(Fmoc)-OH

Purity: 95% CAS No.: 117106-21-5 Molecular Formula: C24H28N2O6 Formula Weight: 440.50 Beilstein Reference: 4771583 MDL No.: MFCD00236845

Formel: C₂₄H₂₈N₂O₆
Molecular Weight: 440,496 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236845
CAS-Nummer: 117106-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR