(S)-3-Amino-5-hexynoic acid hydrochloride 95%

Lieferant: Thermo Fisher Scientific
Achtung

H52010.MD H52010.03
H52010.MDEA 194 EUR
H52010.MD H52010.03
(S)-3-Amino-5-hexynoic acid hydrochloride 95%
(S)-3-Amino-5-hexynoic acid hydrochloride

Purity: 95% CAS No.: 270596-46-8 Molecular Formula: C6H9NO2.HCl Formula Weight: 163.60 MDL No.: MFCD01861006

Formel: C₆H₉NO₂·HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD01861006
CAS-Nummer: 270596-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR