Boc-3-chloro-L-phenylalanine 95%

Lieferant: Thermo Fisher Scientific

Synonyme: 3-Chloro-L-phenylalanine, N-BOC protected, Boc-Phe(3-Cl)-OH

H52015.03 H52015.06
H52015.03EA 77 EUR
H52015.03 H52015.06
Boc-3-chloro-L-phenylalanine 95%
Boc-3-chloro-L-phenylalanine

Purity: 95% CAS No.: 114873-03-9 Molecular Formula: C14H18ClNO4 Formula Weight: 299.75 MDL No.: MFCD00672515

Formel: C₁₄H₁₈ClNO₄
Molecular Weight: 299.76 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672515
CAS-Nummer: 114873-03-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR