(S)-3-Amino-3-(3-nitrophenyl)propionic acid 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(S)-3-Amino-3-(3-nitrophenyl)propanoic acid
H52027.03
H52027.06
H52027.03EA
184
EUR
H52027.03
H52027.06
(S)-3-Amino-3-(3-nitrophenyl)propionic acid 95%
(S)-3-Amino-3-(3-nitrophenyl)propionic acid
Purity: 95%
CAS No.: 734529-57-8
Molecular Formula: C9H10N2O4
Formula Weight: 210.19
MDL No.: MFCD04113693
Formel:
C₉H₁₀N₂O₄ Molecular Weight: 210,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04113693 CAS-Nummer: 734529-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...