Fmoc-4-(boc-amino)-L-phenylalanine 95%

Lieferant: Thermo Scientific

Synonyme: Fmoc-Phe(4-NHBoc)-OH, Fmoc-4-(Boc-amino)-L-phenylalanine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-((tert-butoxycarbonyl)amino)phenyl)propanoic acid

H52031.MD H52031.03
H52031.MDEA 84.1 EUR
H52031.MD H52031.03
Fmoc-4-(boc-amino)-L-phenylalanine 95%
Fmoc-4-(boc-amino)-L-phenylalanine

Purity: 95% CAS No.: 174132-31-1 Molecular Formula: C29H30N2O6 Formula Weight: 502.57 MDL No.: MFCD00237007

Formel: C₂₉H₃₀N₂O₆
Molecular Weight: 502,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00237007
CAS-Nummer: 174132-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR