Fmoc-L-β-homoproline 95%
Lieferant: Thermo Scientific
Synonyme:
Fmoc-β-HomoPro-OH, Fmoc-β-HoPro-OH
H52060.MD
H52060.03
H52060.MDEA
117
EUR
H52060.MD
H52060.03
Fmoc-L-β-homoproline 95%
Fmoc-L-beta-homoproline
Purity: 95%
CAS No.: 193693-60-6
Molecular Formula: C21H21NO4
Formula Weight: 351.40
Beilstein Reference: 7798553
MDL No.: MFCD01863058
Formel:
C₂₁H₂₁NO₄ Molecular Weight: 351,41 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863058 CAS-Nummer: 193693-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...