Fmoc-O-t-butyl-L-β-homoserine 95%
Lieferant: Thermo Scientific
Synonyme:
Fmoc-β-HomoSer(tBu)-OH, Fmoc-β-HoSer(tBu)-OH
H52065.03
H52065.MD
H52065.03EA
443
EUR
H52065.03
H52065.MD
Fmoc-O-t-butyl-L-β-homoserine 95%
Fmoc-O-t-butyl-L-beta-homoserine
Purity: 95%
CAS No.: 203854-51-7
Molecular Formula: C23H27NO5
Formula Weight: 397.47
Melting Point: 96-98°
Beilstein Reference: 8014825
MDL No.: MFCD01862865
Formel:
C₂₃H₂₇NO₅ Molecular Weight: 397,48 g/mol Schmelzpunkt: 96…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862865 CAS-Nummer: 203854-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...