N-Fmoc-3-(4-thiazolyl)-L-alanine 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-3-(4-thiazolyl)-Alanine, Fmoc-β-(4-Thiazolyl)-Ala-OH, Fmoc-L-(4-thiazolyl)-Alanine
H52071.MD
H52071.03
H52071.06
H52071.MDEA
67.8
EUR
H52071.MD
H52071.03
H52071.06
N-Fmoc-3-(4-thiazolyl)-L-alanine 95%
N-Fmoc-3-(4-thiazolyl)-L-alanine
Purity: 95%
CAS No.: 205528-32-1
Molecular Formula: C21H18N2O4S
Formula Weight: 394.44
Melting Point: 178-179°
MDL No.: MFCD00672568
Formel:
C₂₁H₁₈N₂O₄S Molecular Weight: 394.4 g/mol Schmelzpunkt: 178…179 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672568 CAS-Nummer: 205528-32-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...