Fmoc-L-4-methylphe 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-methyl-L-phenylalanine, N-Fmoc-4-methyl-L-phenylalanine, Fmoc-Phe(4-Me)-OH, Fmoc-p-Me-Phe-OH
H52073.03
H52073.06
H52073.03EA
102
EUR
H52073.03
H52073.06
Fmoc-L-4-methylphe 95%
Fmoc-L-4-methylphe
Purity: 95%
CAS No.: 199006-54-7
Molecular Formula: C25H23NO4
Formula Weight: 401.46
Beilstein Reference: 8799411
MDL No.: MFCD00270197
Formel:
C₂₅H₂₃NO₄ Molecular Weight: 401.462 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00270197 CAS-Nummer: 199006-54-7 |