L-4-Benzoylphenylalanine 95%

Lieferant: Thermo Fisher Scientific

Synonyme: (S)-2-Amino-3-(4-benzoylphenyl)propanoic acid, H-Bpa-OH, H-p-Bz-Phe-OH, H-Phe(4-Bz)-OH

H52083.MD H52083.03 H52083.06
H52083.MDEA 108 EUR
H52083.MD H52083.03 H52083.06
L-4-Benzoylphenylalanine 95%
L-4-Benzoylphenylalanine

Purity: 95% CAS No.: 104504-45-2 Molecular Formula: C16H15NO3 Formula Weight: 269.30 MDL No.: MFCD00238102

Formel: C₁₆H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00238102
CAS-Nummer: 104504-45-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR