Boc-D-allyl-Gly-OH dicyclohexylammonium salt 95%

Lieferant: Thermo Scientific

Synonyme: Boc-D-allylglycine (dicyclohexylammonium) salt, 2-Allyl-N-Boc-D-glycine dicyclohexylamine salt, Boc-D-Gly(Allyl)-OH·DCHA, D-2-Allylglycine, N-BOC protected dicylohexylamine salt, Boc-D-allylglycine.DCHA, Dicyclohexylamine (R)-2-((tert-butoxycarbonyl)amino)pent-4-enoate, (R)-2-(Boc-amino)-4-pentenoic acid (dicyclohexylammonium) salt

H52127.03 H52127.06
H52127.03EA 75.2 EUR
H52127.03 H52127.06
Boc-D-allyl-Gly-OH dicyclohexylammonium salt 95%
Boc-D-allyl-Gly-OH dicyclohexylammonium salt

Purity: 95% CAS No.: 170899-08-8 Molecular Formula: C10H17NO4.C12H23N Formula Weight: 396.57 MDL No.: MFCD01321012

Formel: C10H17NO4.C12H23N
Molecular Weight: 396,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01321012
CAS-Nummer: 170899-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR