Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 95%

Lieferant: Thermo Fisher Scientific

Synonyme: 3-Thien-3-yl-L-alanine, N-FMOC protected

H52167.MD H52167.03
H52167.MDEA 87.1 EUR
H52167.MD H52167.03
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 95%
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH

Purity: 95% CAS No.: 186320-06-9 Molecular Formula: C22H19NO4S Formula Weight: 393.46 Beilstein Reference: 9512976 MDL No.: MFCD00151916

Formel: C₂₂H₁₉NO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00151916
CAS-Nummer: 186320-06-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR