Fmoc-L-4-pyridylalanine 95%
Lieferant: Thermo Scientific
Synonyme:
N-Fmoc-3-(4-pyridyl)-L-alanine, Fmoc-3-(4-pyridyl)-Ala-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(4-pyridyl)-L-alanine, Fmoc-4-Pal-OH
H52172.MD
H52172.03
H52172.06
H52172.MDEA
80.8
EUR
H52172.MD
H52172.03
H52172.06
Fmoc-L-4-pyridylalanine 95%
Fmoc-L-4-pyridylalanine
Purity: 95%
CAS No.: 169555-95-7
Molecular Formula: C23H20N2O4
Formula Weight: 388.42
Beilstein Reference: 7316905
MDL No.: MFCD00672566
Formel:
C₂₃H₂₀N₂O₄ Molecular Weight: 388,43 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672566 CAS-Nummer: 169555-95-7 |