Boc-L-β-homoleucine 95%
Lieferant: Thermo Scientific
Synonyme:
Boc-β-HoLeu-OH, (S)-3-((tert-Butoxycarbonyl)amino)-5-methylhexanoic acid
H52173.MD
H52173.03
H52173.MDEA
116
EUR
H52173.MD
H52173.03
Boc-L-β-homoleucine 95%
Boc-L-beta-homoleucine
Purity: 95%
CAS No.: 132549-43-0
Molecular Formula: C12H23NO4
Formula Weight: 245.32
Melting Point: 53-55°
Beilstein Reference: 4251252
MDL No.: MFCD02101665
Formel:
C₁₂H₂₃NO₄ Molecular Weight: 245,32 g/mol Schmelzpunkt: 53…55 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02101665 CAS-Nummer: 132549-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...