Fmoc-L-β-homovaline 95%
Lieferant: Thermo Scientific
Synonyme:
Fmoc-β-HoVal-OH, Fmoc-β-HomoVal-OH
H52182.MD
H52182.03
H52182.MDEA
118
EUR
H52182.MD
H52182.03
Fmoc-L-β-homovaline 95%
Fmoc-L-beta-homovaline
Purity: 95%
CAS No.: 172695-33-9
Molecular Formula: C21H23NO4
Formula Weight: 353.42
Beilstein Reference: 7388393
MDL No.: MFCD01862853
Formel:
C₂₁H₂₃NO₄ Molecular Weight: 353,42 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862853 CAS-Nummer: 172695-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...