Fmoc-L-β-homoaspartic acid-gamma-t-butyl ester 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-β-HomoAsp(OtBu)-OH, Fmoc-β-HoAsp(OtBu)-OH
H52190.MD
H52190.03
H52190.MDEA
155
EUR
H52190.MD
H52190.03
Fmoc-L-β-homoaspartic acid-gamma-t-butyl ester 95%
Fmoc-L-beta-homoaspartic acid-gamma-t-butyl ester
Purity: 95%
CAS No.: 209252-17-5
Molecular Formula: C24H27NO6
Formula Weight: 425.48
Beilstein Reference: 7952342
MDL No.: MFCD01862860
Formel:
C₂₄H₂₇NO₆ Molecular Weight: 425.49 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862860 CAS-Nummer: 209252-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...