Fmoc-O-t-butyl-L-β-homotyrosine 95%

Lieferant: Thermo Fisher Scientific

Synonyme: Fmoc-β-HoTyr(tBu)-OH, Fmoc-β-HomoTyr(tBu)-OH

H52192.MD H52192.03
H52192.MDEA 155 EUR
H52192.MD H52192.03
Fmoc-O-t-butyl-L-β-homotyrosine 95%
Fmoc-O-t-butyl-L-beta-homotyrosine

Purity: 95% CAS No.: 219967-69-8 Molecular Formula: C29H31NO5 Formula Weight: 473.57 Beilstein Reference: 8236890 MDL No.: MFCD01862862

Formel: C₂₉H₃₁NO₅
Molecular Weight: 473.57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01862862
CAS-Nummer: 219967-69-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR