1-(2-Pyridinyl)-1-ethanone oxime 97%
Lieferant: Thermo Fisher Scientific
Achtung
H52232.03 H52232.06
H52232.03EA 45.9 EUR
H52232.03 H52232.06
1-(2-Pyridinyl)-1-ethanone oxime 97%
1-(2-Pyridinyl)-1-ethanone oxime
Purity: 97% CAS No.: 1758-54-9 Molecular Formula: C7H8N2O Formula Weight: 136.15 MDL No.: MFCD00605540
Formel: C₇H₈N₂O
Molecular Weight: 136.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00499500
CAS-Nummer: 1758-54-9