3-(Boc-amino)cyclohexanecarboxylic acid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid
H52299.03
H52299.06
H52299.03EA
111
EUR
H52299.03
H52299.06
3-(Boc-amino)cyclohexanecarboxylic acid ≥98%
3-(Boc-amino)cyclohexanecarboxylic acid
Purity: 98+%
CAS No.: 334932-13-7
Molecular Formula: C12H21NO4
Formula Weight: 243.30
MDL No.: MFCD03453252
Formel:
C₁₂H₂₁NO₄ Molecular Weight: 243,30 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03453252 CAS-Nummer: 334932-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...