(S)-(-)-2-Amino-3-benzyloxy-1-propanol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
H-D-Ser(Bzl)-ol
H52304.03
H52304.06
H52304.03EA
87.8
EUR
H52304.03
H52304.06
(S)-(-)-2-Amino-3-benzyloxy-1-propanol ≥98%
(S)-(-)-2-Amino-3-benzyloxy-1-propanol
Purity: 98+%
CAS No.: 58577-88-1
Molecular Formula: C10H15NO2
Formula Weight: 181.23
UN No.: UN3267
MDL No.: MFCD04112477
Formel:
C₁₀H₁₅NO₂ Molecular Weight: 181,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04112477 CAS-Nummer: 58577-88-1 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...