Boc-5-amino-2-chlorobenzoic acid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H52308.03 H52308.06
H52308.03EA 192 EUR
H52308.03 H52308.06
Boc-5-amino-2-chlorobenzoic acid ≥98%
Boc-5-amino-2-chlorobenzoic acid

Purity: 98+% CAS No.: 503555-96-2 Molecular Formula: C12H14ClNO4 Formula Weight: 271.70 MDL No.: MFCD02682166

Formel: C₁₂H₁₄ClNO₄
Molecular Weight: 271.7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02682166
CAS-Nummer: 503555-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR