5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoindolin-1-one 96%
Lieferant: Thermo Fisher Scientific
Synonyme: 1-Oxoisoindoline-5-boronic acid, pinacol ester
H52317.MD H52317.03
H52317.MDEA 190 EUR
H52317.MD H52317.03
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoindolin-1-one 96%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoindolin-1-one
Purity: 96% CAS No.: 376584-62-2 Molecular Formula: C14H18BNO3 Formula Weight: 259.11 MDL No.: MFCD11040393
Formel: C₁₄H₁₈BNO₃
Molecular Weight: 259.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11040393
CAS-Nummer: 376584-62-2
Bestellen
TECHNISCHE DATEN
Environmentally Preferable