7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥97%

Lieferant: Thermo Scientific
Achtung

H52389.03 H52389.06
H52389.03EA 213 EUR
H52389.03 H52389.06
7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥97%
7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride

Purity: 97+% CAS No.: 99365-69-2 Molecular Formula: C9H10N2O2.HCl Formula Weight: 214.65 MDL No.: MFCD07371494

Formel: C₉H₁₁ClN₂O₂
Molecular Weight: 214,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371494
CAS-Nummer: 99365-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR