Ethyl-4-methyl-1,3-oxazol-5-carboxylat ≥97%
Lieferant: Thermo Scientific
Synonyme:
Ethyl 4-methyloxazole-5-carboxylate
H52404.03
H52404.06
H52404.03EA
131
EUR
H52404.03
H52404.06
Ethyl-4-methyl-1,3-oxazol-5-carboxylat ≥97%
Ethyl-4-methyloxazole-5-carboxylate
Purity: 97+%
CAS No.: 20485-39-6
Molecular Formula: C6H7NO3
Formula Weight: 141.10
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00062573
Formel:
C₇H₉NO₃ Molecular Weight: 155,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00062573 CAS-Nummer: 20485-39-6 EINECS: 243-848-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...