Boc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid 97%
Lieferant: Thermo Scientific
Synonyme:
Boc-Val(3-OH)-OH, 3-Hydroxy-L-valine, N-BOC protected, (S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxy-3-methylbutanoic acid
H52429.MD
H52429.03
H52429.MDEA
175
EUR
H52429.MD
H52429.03
Boc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid 97%
Boc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid
Purity: 97%
CAS No.: 102507-13-1
Molecular Formula: C10H19NO5
Formula Weight: 233.26
MDL No.: MFCD03094792
Formel:
C₁₀H₁₉NO₅ Molecular Weight: 233,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094792 CAS-Nummer: 102507-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...