Boc-Lys(ME)2-OH 97%

Lieferant: Thermo Fisher Scientific

Synonyme: Boc-Lys(Me)₂-OH

H52437.MD H52437.03
H52437.MDEA 87 EUR
H52437.MD H52437.03
Boc-Lys(ME)2-OH 97%
Boc-Lys(ME)2-OH

Purity: 97% CAS No.: 65671-53-6 Molecular Formula: C13H26N2O4 Formula Weight: 274.36 MDL No.: MFCD00076965

Formel: C₁₃H₂₆N₂O₄
Molecular Weight: 274.361 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00076965
CAS-Nummer: 65671-53-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR