3-(2-Methoxycarbonylethyl)benzeneboronic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (3-(3-Methoxy-3-oxopropyl)phenyl)boronic acid

H52542.MD H52542.03
H52542.MDEA 81.5 EUR
H52542.MD H52542.03
3-(2-Methoxycarbonylethyl)benzeneboronic acid 97%
3-(2-Methoxycarbonylethyl)benzeneboronic acid

Purity: 97% CAS No.: 833472-82-5 Molecular Formula: C10H13BO4 Formula Weight: 208.02 Melting Point: 96-100° MDL No.: MFCD04115648

Formel: C₁₀H₁₃BO₄
Molecular Weight: 208.02 g/mol
Schmelzpunkt: 96…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115648
CAS-Nummer: 833472-82-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR