9-Phenanthreneboronic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H52546.03
H52546.06
H52546.03EA
37.7
EUR
H52546.03
H52546.06
9-Phenanthreneboronic acid 97%
9-Phenanthreneboronic acid
Purity: 97%
CAS No.: 68572-87-2
Molecular Formula: C14H11BO2
Formula Weight: 222.05
Melting Point: 254-256°
MDL No.: MFCD00143524
Formel:
C₁₄H₁₁BO₂ Molecular Weight: 222.05 g/mol Schmelzpunkt: 254…256 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143524 CAS-Nummer: 68572-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...