Boc-L-α-methylserine 97%

Lieferant: Thermo Scientific

H52570.MD H52570.03
H52570.MDEA 120 EUR
H52570.MD H52570.03
Boc-L-α-methylserine 97%
Boc-L-alpha-methylserine

Purity: 97% CAS No.: 84311-19-3 Molecular Formula: C9H17NO5 Formula Weight: 219.24 MDL No.: MFCD02682593

Formel: C₉H₁₇NO₅
Molecular Weight: 219,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02682593
CAS-Nummer: 84311-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR