4-(Acetamidomethyl)benzeneboronic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-[(Acetylamino)methyl]benzeneboronic acid, (4-(Acetamidomethyl)phenyl)boronic acid

H52602.MD H52602.03
H52602.MDEA 82.7 EUR
H52602.MD H52602.03
4-(Acetamidomethyl)benzeneboronic acid 97%
4-(Acetamidomethyl)benzeneboronic acid

Purity: 97% CAS No.: 850568-41-1 Molecular Formula: C9H12BNO3 Formula Weight: 193.01 Melting Point: 278-280° MDL No.: MFCD06659818

Formel: C₉H₁₂BNO₃
Molecular Weight: 193.01 g/mol
Schmelzpunkt: 278…280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659818
CAS-Nummer: 850568-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR