(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Amino-2-(hydroxymethyl)benzeneboronic acid dehydrate hydrochloride, 6-Aminobenzo[c][1,2]oxaborol-1(3H)-ol hydrochloride

H52629.MD H52629.03
H52629.MDEA 185 EUR
H52629.MD H52629.03
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride 95%
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride

Purity: 95% CAS No.: 117098-93-8 Molecular Formula: C7H8BNO2.HCl Formula Weight: 185.42 Melting Point: 300° MDL No.: MFCD04115645

Formel: C₇H₉BClNO₂
Molecular Weight: 185,42 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115645
CAS-Nummer: 117098-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR