(S)-1-Boc-2-(aminomethyl)pyrrolidine 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (S)-2-(Aminomethyl)-1-Boc-pyrrolidine, (S)-2-Aminomethyl-1-Boc-pyrrolidine, tert-Butyl (S)-2-(aminomethyl)-1-pyrrolidinecarboxylate, (S)-1-Boc-2-(Aminomethyl)pyrrolidine, (2S)-2-(Aminomethyl)pyrrolidine, N1-BOC protected, (S)-2-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine, (S)-2-(Aminomethyl)-1-pyrrolidinecarboxylic acid tert-Butyl Ester

H52727.03 H52727.06
H52727.03EA 89.3 EUR
H52727.03 H52727.06
(S)-1-Boc-2-(aminomethyl)pyrrolidine 97%
(S)-1-Boc-2-(aminomethyl)pyrrolidine

Purity: 97% CAS No.: 119020-01-8 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 Boiling Point: 98-112°/1mm UN No.: UN2735 MDL No.: MFCD03419257

Formel: C₁₀H₂₀N₂O₂
Siedepunkt: 98…112 °C (1 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD03419257
CAS-Nummer: 119020-01-8
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR