Fmoc-Hyp-OH 97%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Fmoc-trans-4-hydroxy-L-proline

H52740.03 H52740.06
H52740.03EA 35.1 EUR
H52740.03 H52740.06
Fmoc-Hyp-OH 97%
Fmoc-Hyp-OH

Purity: 97% CAS No.: 88050-17-3 Molecular Formula: C20H19NO5 Formula Weight: 353.37 Melting Point: 189-193° Optical Rotation: -60° (c=1 in methanol) MDL No.: MFCD00151929

Formel: C₂₀H₁₉NO₅
Molecular Weight: 353.374 g/mol
Schmelzpunkt: 189…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151929
CAS-Nummer: 88050-17-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR