(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid 97%

Lieferant: Thermo Scientific

H52775.MD H52775.03
H52775.MDEA 224 EUR
H52775.MD H52775.03
(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid 97%
(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid

Purity: 97% CAS No.: 62023-65-8 Molecular Formula: C15H21NO5 Formula Weight: 295.34 Beilstein Reference: 5909790 MDL No.: MFCD00057841

Formel: C₁₅H₂₁NO₅
Molecular Weight: 295,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00057841
CAS-Nummer: 62023-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR