(1S,2S)-2-Aminocyclohexancarbonsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

H52779.MD H52779.03 H52779.06
H52779.MDEA 116 EUR
H52779.MD H52779.03 H52779.06
(1S,2S)-2-Aminocyclohexancarbonsäure 97%
(1S,2S)-2-Aminocyclohexanecarboxylic acid

Purity: 97% CAS No.: 24716-93-6 Molecular Formula: C7H13NO2 Formula Weight: 143.19 Optical Rotation: +77° (c=1 in water) MDL No.: MFCD01318270

Formel: C₇H₁₃NO₂
Molecular Weight: 143.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01318270
CAS-Nummer: 24716-93-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR