Boc-4-amino-2-chlorobenzoic acid 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-((tert-Butoxycarbonyl)amino)-2-chlorobenzoic acid
H52786.03
H52786.06
H52786.03EA
157
EUR
H52786.03
H52786.06
Boc-4-amino-2-chlorobenzoic acid 97%
Boc-4-amino-2-chlorobenzoic acid
Purity: 97%
CAS No.: 232275-73-9
Molecular Formula: C12H14ClNO4
Formula Weight: 271.70
MDL No.: MFCD02682164
Formel:
C₁₂H₁₄ClNO₄ Molecular Weight: 271,7 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682164 CAS-Nummer: 232275-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...