Lovastatin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6alpha-Methylcompactin, Monacolin K, Mevinolin, (S)-(1S,3R,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2-methylbutanoate

H52792.06 H52792.14
H52792.06EA 91.9 EUR
H52792.06 H52792.14
Lovastatin 97%
Lovastatin

Purity: 97% CAS No.: 75330-75-5 Molecular Formula: C24H36O5 Formula Weight: 404.56 Melting Point: 175° MDL No.: MFCD00072164

Formel: C₂₄H₃₆O₅
Molecular Weight: 404,55 g/mol
Schmelzpunkt: 175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00072164
CAS-Nummer: 75330-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR