N-(α)-Boc-DL-2,3-diaminopropionic acid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Amino-2-((tert-butoxycarbonyl)amino)propanoic acid, 3-Amino-N-(tert-butoxycarbonyl)alanine, Boc-DL-α-3-NH₂-Ala-OH

H52811.03 H52811.06
H52811.03EA 52.4 EUR
H52811.03 H52811.06
N-(α)-Boc-DL-2,3-diaminopropionic acid 97%
N-(alpha)-Boc-DL-2,3-diaminopropionic acid

Purity: 97% CAS No.: 159002-17-2 Molecular Formula: C8H16N2O4 Formula Weight: 204.23 Physical Form: Crystalline solid Appearance: White MDL No.: MFCD00736752

Formel: C₈H₁₆N₂O₄
Molecular Weight: 204,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00736752
CAS-Nummer: 159002-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR