2-Methyl-2-propanyl-4-(aminomethyl)-1-piperidincarboxylat 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-(aminomethyl)piperidine-1-carboxylate, tert-Butyl 4-(Aminomethyl)-1-piperidinecarboxylate, tert-Butyl 4-(aminomethyl)tetrahydropyridine-1(2H)- carboxylate, 4-(Aminomethyl)-1-Boc-piperidine, 4-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-4-(Aminomethyl)piperidine

H52819.06 H52819.14
H52819.06EA 61.6 EUR
H52819.06 H52819.14
2-Methyl-2-propanyl-4-(aminomethyl)-1-piperidincarboxylat 97%
1-Boc-4-(aminomethyl)piperidine

Purity: 97% CAS No.: 144222-22-0 Molecular Formula: C11H22N2O2 Formula Weight: 214.31 Flash Point: 110°(230°F) Boiling Point: 237-238° MDL No.: MFCD01076207

Formel: C₁₁H₂₂N₂O₂
Molecular Weight: 214.31 g/mol
Siedepunkt: 237…238 °C
Dichte: 1.013
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD01076207
CAS-Nummer: 144222-22-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR