(R)-3-Amino-2-((tert-butoxycarbonyl)amino)propanoic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Boc-D-Dap-OH
H52821.MD
H52821.03
H52821.06
H52821.MDEA
59.6
EUR
H52821.MD
H52821.03
H52821.06
(R)-3-Amino-2-((tert-butoxycarbonyl)amino)propanoic acid 97%
(R)-3-Amino-2-((tert-butoxycarbonyl)amino)propanoic acid
Purity: 97%
CAS No.: 76387-70-7
Molecular Formula: C8H16N2O4
Formula Weight: 204.23
Melting Point: 200-203°
Optical Rotation: +14° (c=1 in water)
MDL No.: MFCD01632072
Formel:
C₈H₁₆N₂O₄ Schmelzpunkt: 200…203 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01632072 CAS-Nummer: 76387-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...