Fmoc-4-Abz-OH 97%

Lieferant: Thermo Fisher Scientific

H52828.MD H52828.03
H52828.MDEA 49.3 EUR
H52828.MD H52828.03
Fmoc-4-Abz-OH 97%
Fmoc-4-Abz-OH

Purity: 97% CAS No.: 185116-43-2 Molecular Formula: C22H17NO4 Formula Weight: 359.38 Beilstein Reference: 7722986 MDL No.: MFCD00144888

Formel: C₂₂H₁₇NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00144888
CAS-Nummer: 185116-43-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR